Skip to Content
Merck
All Photos(1)

Key Documents

218707

Sigma-Aldrich

Cyclophosphamide monohydrate

≥98%

Synonym(s):

2-[Bis(2-chloroethyl)amino]tetrahydro-2H-1,3,2-oxazaphosphorine 2-oxide, Cytoxan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15Cl2N2O2P · H2O
CAS Number:
Molecular Weight:
279.10
Beilstein:
4678992
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

≥98%

mp

49-51 °C (lit.)

SMILES string

[H]O[H].ClCCN(CCCl)P1(=O)NCCCO1

InChI

1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2

InChI key

PWOQRKCAHTVFLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Cyclophosphamide is a cytotoxic nitrogen mustard derivative widely used in cancer chemotherapy. It cross-links DNA, causes strand breakage, and induces mutations. Its clinical activity is associated with a decrease in aldehyde dehydrogenase 1 (ALDH1) activity.
Cytotoxic nitrogen mustard derivative widely used in cancer chemotherapy. It cross-links DNA, causes strand breakage, and induces mutations.

Features and Benefits

Cancer research tool.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Bastaki et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 105, 278-284 (2017-04-30)
Tartrazine is approved as a food color additive internationally with INS number 102, in the United States as food color subject to batch certification "Food, Drug, and Cosmetic" (FD&C) Yellow No. 5, and in Europe as food color additive with
Karen Gomez-Mariscal et al.
Annals of clinical and laboratory science, 48(2), 152-157 (2018-04-22)
The aim of the study was to determine if amniotic fluid cells of rats can be used to provide evidence of genotoxicity. In order to do that micronuclei formation was induced in rats during pregnancy after treatment with cyclophosphamide (CP)
Marie Z Vasquez
Mutagenesis, 25(2), 187-199 (2009-12-09)
Despite regulatory directives requiring the reduction of animal use in safety testing, recent modifications to genotoxicity testing guidelines now propose the use of two in vivo genotoxicity assays as a follow-up to an in vitro positive (International Conference on Harmonization
Ricardo I Peraza-Vega et al.
Toxicology and industrial health, 33(5), 443-453 (2016-10-26)
The aim of this study was to evaluate the genotoxicity of the herbicide diuron in the wing-spot test and a novel wing imaginal disk comet assay in Drosophila melanogaster. The wing-spot test was performed with standard (ST) and high-bioactivation (HB)
S Khan et al.
Toxicology international, 20(1), 68-76 (2013-07-09)
Recently, sodium valproate (VPA) has been proven as histone deacetylase (HDAC) inhibitor and potentiates the cytotoxicity of anticancer drugs, and also exhibit promising anti-cancer activity. Present study aimed to investigate the influence of pre- and post-treatment of VPA on cyclophosphamide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service