Skip to Content
Merck
All Photos(1)

Key Documents

239785

Sigma-Aldrich

Cyclophosphamide Monohydrate

≥98% (HPLC), solid, anti-neoplastic agent, Calbiochem®

Synonym(s):

Cyclophosphamide Monohydrate, N,N- bis(2-Chloroethyl)tetrahydro-2H-1,3,2-oxazaphosphorin-2-amine-2-oxide Monohydrate, CPA, N,N-bis(2-Chloroethyl)tetrahydro-2H-1,3,2-oxazaphosphorin-2-amine-2-oxide Monohydrate, CPA

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€143.00

€143.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€143.00

About This Item

Empirical Formula (Hill Notation):
C7H15Cl2N2O2P · xH2O
CAS Number:
Molecular Weight:
261.09 (anhydrous basis)
MDL number:
UNSPSC Code:
51112507
NACRES:
NA.77

€143.00


Estimated to ship on30 May 2025


Request a Bulk Order

Product Name

Cyclophosphamide Monohydrate, A widely used anti-neoplastic agent that also displays immunosuppressant properties.

Quality Level

description

Merck USA index - 14, 747

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 30 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[P]1(=O)(NCCCO1)N(CCCl)CCCl.O

InChI

1S/C7H15Cl2N2O2P.H2O/c8-2-5-11(6-3-9)14(12)10-4-1-7-13-14;/h1-7H2,(H,10,12);1H2

InChI key

PWOQRKCAHTVFLB-UHFFFAOYSA-N

General description

A widely used anti-neoplastic agent that also displays immunosuppressant properties. Induces apoptotic cell death in rat thymus through the Fas/Fas ligand pathway.
A widely used antineoplastic agent that also displays immuno-suppressant activity. Induces apoptotic cell death in rat thymus throught Fas/Fas ligand pathway.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Fas/Fas ligand pathway
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Toxic & Carcinogenic / Teratogenic (G)

Reconstitution

Solutions should be prepared fresh on the day of use.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Wang, G.J., and Cai, L. 1999. Mutat. Res.427, 125.
Lee, J.H., et al. 1997. Korean J. Med. Sci. 12, 185.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1B - Muta. 1B - Repr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service