Skip to Content
Merck
All Photos(1)

Key Documents

214663

Sigma-Aldrich

2-Ethylanthracene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14
CAS Number:
Molecular Weight:
206.28
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

form

solid

mp

152-153 °C (lit.)

SMILES string

CCc1ccc2cc3ccccc3cc2c1

InChI

1S/C16H14/c1-2-12-7-8-15-10-13-5-3-4-6-14(13)11-16(15)9-12/h3-11H,2H2,1H3

InChI key

ZXAGXLDEMUNQSH-UHFFFAOYSA-N

General description

Photophysics and photochemistry of 2-ethylanthracene (2EA) adsorbed on silica has been studied at a silica/air interface.

Application

2-Ethylanthracene was used as photosensitizer during the photodissociation of p-nitrobenzyl esters of alkylsulphonic acids. It was used in the synthesis of thioxanthone-2-ethyl anthracene.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thioxanthone-ethyl anthracene.
Balta D-K and Arsu N.
Journal of Photochemistry and Photobiology A: Chemistry, 257, 54-59 (2013)
Photophysical and photochemical processes of 2-methyl, 2-ethyl, and 2-tert-butylanthracenes on silica gel. A substituent effect study.
Dabestani R, et al.
The Journal of Physical Chemistry B, 104(44), 10235-10241 (2000)
Peter A Beckmann et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 19(18), 2423-2436 (2018-06-30)
We report solid-state nuclear magnetic resonance 1 H spin-lattice relaxation, single-crystal X-ray diffraction, powder X-ray diffraction, field emission scanning electron microscopy, and differential scanning calorimetry in solid samples of 2-ethylanthracene (EA) and 2-ethylanthraquinone (EAQ) that have been physically purified in
Photochemical dissociation of p-nitrobenzyl 9, 10-dimethoxyanthracene-2-sulphonate via intramolecular electron transfer.
Yamaoka T, et al.
J. Chem. Soc. Perkin Trans. II, 10, 1709-1714 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service