Skip to Content
Merck
All Photos(2)

Documents

208949

Sigma-Aldrich

2-Fluorophenylacetic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CO2H
CAS Number:
Molecular Weight:
154.14
Beilstein:
775955
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

60-62 °C (lit.)

SMILES string

OC(=O)Cc1ccccc1F

InChI

1S/C8H7FO2/c9-7-4-2-1-3-6(7)5-8(10)11/h1-4H,5H2,(H,10,11)

InChI key

RPTRFSADOICSSK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Fluorophenylacetic acid is chiral derivatizing agent for determination of enantiomeric composition of chiral, nonracemic compounds by 19F NMR spectroscopy.

Application

2-Fluorophenylacetic acid was used:
  • in the synthesis of thiazolino[3,2-c]pyrimidin-5,7-diones and N-[2-(3,4-dichlorophenyl)-ethyl]-N′-[2-(2-fluorophenyl)-ethyl]-ethane-1,2-diamine
  • as chiral derivatizing agent for determination of enantiomeric composition of chiral, nonracemic compounds by 19F NMR spectroscopy

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boron trifluoride-induced reactions of phenylglycidyl ethers: a convenient synthesis of enantiopure, stereodefined fluorohydrins.
Islas-Gonzalez G, et al.
Tetrahedron Letters, 45(33), 6337-6341 (2004)
Joseph Pontillo et al.
Bioorganic & medicinal chemistry letters, 15(5), 1407-1411 (2005-02-17)
Treatment of various 2-methyl oxazolines or thiazolines with chlorocarbonyl isocyanate gives the corresponding bicyclic oxazolino- or thiazolino[3,2-c]pyrimidin-5,7-dione derivatives in very good yield. This reaction has been applied to the rapid syntheses of human gonadotropin-releasing hormone (hGnRH) receptor antagonists for SAR
Yaqun Li et al.
The Korean journal of pain, 34(1), 27-34 (2021-01-01)
Chemotherapy-induced peripheral neuropathy (CIPN) is a major reason for stopping or changing anticancer therapy. Among the proposed pathomechanisms underlying CIPN, proinflammatory processes have attracted increasing attention. Here we assessed the role of prostaglandin D2 (PGD2) signaling in cisplatin-induced neuropathic pain.
Jonathan M Fitzsimmons et al.
Medicinal chemistry, 1(102) (2011-12-25)
Eight halogenated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service