194336
5-Aminovaleric acid hydrochloride
99%
Synonym(s):
5-Aminopentanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
99%
reaction suitability
reaction type: solution phase peptide synthesis
mp
95-97 °C (lit.)
application(s)
peptide synthesis
SMILES string
Cl[H].NCCCCC(O)=O
InChI
1S/C5H11NO2.ClH/c6-4-2-1-3-5(7)8;/h1-4,6H2,(H,7,8);1H
InChI key
BLOIUFYKQCCAGP-UHFFFAOYSA-N
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Langmuir : the ACS journal of surfaces and colloids, 26(18), 14799-14805 (2010-08-28)
We report the evidence for the cellular uptake of gold nanoparticles via the phagocytosis mechanism in murine macrophage cells strongly supported by TEM and optical microscopy. Nanoparticles were prepared using several biocompatible molecules of choice (5-aminovaleric acid, l-DOPA, melatonin, and
International journal of oral and maxillofacial surgery, 39(8), 802-805 (2010-04-23)
Photodynamic diagnosis (PDD) is a form of cancer detection based on the administration of an exogenous photo-activated compound that accumulates in malignant cells, followed by appropriate photo irradiation. The authors describe a spectroscopic study of 5-aminolevulinic acid (5-ALA)-generated photosensitizer protoporphyrin
Onkologie, 33(11), 613-615 (2010-10-27)
Basal cell carcinoma (BCC) in the nipple area is extremely rare and can be mistaken for Paget's disease. A 47-year-old woman complained of a slowly growing, scaly erythematous plaque around her right nipple which had developed over several years. Paget's
Bioorganic & medicinal chemistry letters, 18(22), 5975-5977 (2008-08-07)
5-Aminovaleric acid and ornithine were evaluated as linkers for the cyclization of beta-dipeptides. Two linked examples of beta-Ala-beta-Ala were prepared by standard coupling methods and their conformations probed by NMR, CD, and computational means. The data suggest that these non-
Journal of bacteriology, 186(11), 3439-3446 (2004-05-20)
Pseudomonas putida KT2440 is a soil microorganism that attaches to seeds and efficiently colonizes the plant's rhizosphere. Lysine is one of the major compounds in root exudates, and P. putida KT2440 uses this amino acid as a source of carbon
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service