Skip to Content
Merck

Skip To

08339

5-Aminolevulinic acid hydrochloride

≥97.0% (AT)

Synonym(s):

δ-Aminolevulinic acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, 5-Aminolaevulinic acid hydrochloride, ALA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 136,00
5 g
Check Cart for Availability
€ 472,00

About This Item

Linear Formula:
NH2CH2C(O)CH2CH2COOH · HCl
CAS Number:
Molecular Weight:
167.59
UNSPSC Code:
12352209
NACRES:
NA.26
PubChem Substance ID:
EC Number:
226-679-5
Beilstein/REAXYS Number:
3690651
MDL number:

€ 136,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (AT)

form

powder or crystals

impurities

≤2% water

color

white to faint yellow

mp

~150 °C (dec.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

Cl[H].NCC(=O)CCC(O)=O

InChI

1S/C5H9NO3.ClH/c6-3-4(7)1-2-5(8)9;/h1-3,6H2,(H,8,9);1H

InChI key

ZLHFONARZHCSET-UHFFFAOYSA-N

Application

5-Aminolevulinic acid hydrochloride has been used as a supplement for culturing Escherichia coli cells for heme biosynthesis.[1]

Biochem/physiol Actions

Intermediate in heme biosynthesis.
5-Aminolevulinic acid (5-ALA) may interact with several transporters such as glutamate, γ-aminobutyric acid (GABA), di-, tri-peptides, and organic transporters in the blood-brain barrier. It may also act as a precursor for the synthesis of porphyrins in chlorophyll and hemes. 5-ALA exhibits plant growth regulator activities.[2]

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.248021.01543A3785
form

powder or crystals

form

solid

form

powder

form

powder

assay

≥97.0% (AT)

assay

-

assay

99% dry basis (perchloric acid titration)

assay

≥98%

storage temp.

2-8°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

−20°C

mp

~150 °C (dec.)

mp

150-156 °C

mp

>235 °C

mp

150 °C

color

white to faint yellow

color

-

color

-

color

-

impurities

≤2% water

impurities

-

impurities

-

impurities

-


Still not finding the right product?


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters