Skip to Content
Merck
All Photos(1)

Key Documents

179698

Sigma-Aldrich

(−)-2,3:4,6-Di-O-isopropylidene-2-keto-L-gulonic acid monohydrate

98%

Synonym(s):

(−)-Diaceton-2-keto-L-gulonsäure, (−)-Diacetone-2-keto-L-gulonic acid, 2,3:4,6-Di-O-isopropyliden-α-L-xylo-2-hexulofuranosonsäure, 2,3:4,6-Di-O-isopropylidene-α-L-xylo-2-hexulofuranosonic acid, Dikegulac

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€214.00

€214.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
100 G
€214.00

About This Item

Empirical Formula (Hill Notation):
C12H18O7 · H2O
CAS Number:
Molecular Weight:
292.28
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

€214.00


Estimated to ship on01 June 2025


Request a Bulk Order

Assay

98%

optical activity

[α]21/D −20°, c = 2 in methanol

mp

88-90 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]O[H].CC1(C)OC[C@@H]2O[C@]3(OC(C)(C)O[C@H]3[C@@H]2O1)C(O)=O

InChI

1S/C12H18O7.H2O/c1-10(2)15-5-6-7(17-10)8-12(16-6,9(13)14)19-11(3,4)18-8;/h6-8H,5H2,1-4H3,(H,13,14);1H2/t6-,7+,8-,12+;/m0./s1

InChI key

ZFQRGFMVXLSLKZ-QCILGFJPSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service