Skip to Content
Merck
All Photos(1)

Key Documents

17290

Sigma-Aldrich

Fmoc-Lys-OH hydrochloride

≥98.0% (HPLC)

Synonym(s):

Nα-Fmoc-L-lysine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€146.00

€146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€146.00

About This Item

Empirical Formula (Hill Notation):
C21H24N2O4 · HCl
CAS Number:
Molecular Weight:
404.89
Beilstein:
8663370
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

€146.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (HPLC)

form

solid

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

Cl.NCCCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H24N2O4.ClH/c22-12-6-5-11-19(20(24)25)23-21(26)27-13-18-16-9-3-1-7-14(16)15-8-2-4-10-17(15)18;/h1-4,7-10,18-19H,5-6,11-13,22H2,(H,23,26)(H,24,25);1H/t19-;/m0./s1

InChI key

MVMZFAIUUXYFGY-FYZYNONXSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zahraa Youssef et al.
Journal of clinical medicine, 8(12) (2019-12-19)
This study describes the employment of gold nanorods (AuNRs), known for their good reputation in hyperthermia-based cancer therapy, in a hybrid combination of photosensitizers (PS) and peptides (PP). We report here, the design and the synthesis of this nanosystem and
Pia Lazki-Hagenbach et al.
Cells, 10(2) (2021-03-07)
The identification of the Mas-related G-protein-coupled receptors (Mrgpr) as targets of diverse stimuli of mast cells (MCs), including neuropeptides and pseudo-allergy causing drugs, has placed these receptors at a prime position in MC research. However, the species-dependent diversity of these

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service