Skip to Content
Merck
All Photos(2)

Key Documents

171417

Sigma-Aldrich

2-Aminopentane

97%

Synonym(s):

1-Methylbutylamine, 2-Amylamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€53.00

€53.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€53.00

About This Item

Linear Formula:
CH3CH(NH2)CH2CH2CH3
CAS Number:
Molecular Weight:
87.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€53.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.4020 (lit.)

bp

90.5-91.5 °C (lit.)

density

0.736 g/mL at 25 °C (lit.)

SMILES string

CCCC(C)N

InChI

1S/C5H13N/c1-3-4-5(2)6/h5H,3-4,6H2,1-2H3

InChI key

IGEIPFLJVCPEKU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Leipert et al.
Journal of the American Society for Mass Spectrometry, 28(12), 2538-2547 (2017-09-01)
N-acyl homoserine lactones (AHL) are small signal molecules involved in the quorum sensing of many gram-negative bacteria, and play an important role in biofilm formation and pathogenesis. Present analytical methods for identification and quantification of AHL require time-consuming sample preparation
Jan Leipert et al.
Analytical and bioanalytical chemistry, 410(19), 4737-4748 (2018-02-23)
The identification and quantification of molecules involved in bacterial communication are major prerequisites for the understanding of interspecies interactions at the molecular level. We developed a procedure allowing the determination of 2-heptyl-4(1H)-quinolone (HHQ) and 2-heptyl-3-hydroxy-4(1H)-quinolone (PQS) and the virulence factor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service