Skip to Content
Merck
All Photos(1)

Key Documents

296643

Sigma-Aldrich

(R)-(−)-sec-Butylamine

99%

Synonym(s):

(R)-(−)-2-Aminobutane

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€81.20
1 G
€150.00

€81.20


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 MG
€81.20
1 G
€150.00

About This Item

Linear Formula:
C2H5CH(NH2)CH3
CAS Number:
Molecular Weight:
73.14
Beilstein:
1718761
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€81.20


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

optical activity

[α]19/D −7.5°, neat

refractive index

n20/D 1.393 (lit.)

bp

63 °C (lit.)

density

0.72 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CC[C@@H](C)N

InChI

1S/C4H11N/c1-3-4(2)5/h4H,3,5H2,1-2H3/t4-/m1/s1

InChI key

BHRZNVHARXXAHW-SCSAIBSYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(R)-(−)-sec-Butylamine can be used:
  • To prepare (R)-(−)-sec-butylbis(2-hydroxyethyl)amine, an intermediate for the synthesis of aminodiphosphine ligands.[1]
  • As a substrate to demonstrate the stereochemistry of Mitsunobu reaction in the synthesis of secondary amines.[2]
  • As a model compound in the study of singlet-triplet transitions in circular dichroism.[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Stereospecific synthesis of secondary amines by the Mitsunobu reaction
Edwards ML, et al.
Tetrahedron Letters, 31(24), 3417-3420 (1990)
Detection of singlet-triplet transitions in circular dichroism. A study of 2-aminobutane
Basil A and Gedanken A
Journal of the American Chemical Society, 111(26), 9237-9238 (1989)
Synthesis of the New Chiral (R)-and (S)-Aminodiphosphine Ligands sec-Butylbis (2-(diphenylphosphino) ethyl) amine, sec-Butylbis (2-(dicyclohexylphosphino) ethyl) amine, and (alpha-Methylbenzyl) bis (2-(dicyclohexylphosphino) ethyl) amine and Their Organom
Bianchini C, et al.
Organometallics, 16(20), 4403-4414 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service