Skip to Content
Merck
All Photos(1)

Key Documents

164526

Sigma-Aldrich

Dimethyl sulfate-d6

99 atom % D

Synonym(s):

(Methyl sulfate)-d6

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€91.30
10 G
€418.00

€91.30


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€91.30
10 G
€418.00

About This Item

Linear Formula:
(CD3O)2SO2
CAS Number:
Molecular Weight:
132.17
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€91.30


Check Cart for Availability

Request more information

vapor density

4.3 (vs air)

Quality Level

vapor pressure

0.7 mmHg ( 25 °C)

isotopic purity

99 atom % D

Assay

99% (CP)

form

liquid

autoignition temp.

923 °F

technique(s)

protein expression: suitable

refractive index

n20/D 1.385 (lit.)

bp

188 °C (lit.)

mp

-32 °C (lit.)

density

1.397 g/mL at 25 °C

mass shift

M+6

SMILES string

[2H]C([2H])([2H])OS(=O)(=O)OC([2H])([2H])[2H]

InChI

1S/C2H6O4S/c1-5-7(3,4)6-2/h1-2H3/i1D3,2D3

InChI key

VAYGXNSJCAHWJZ-WFGJKAKNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service