Skip to Content
Merck
All Photos(1)

Key Documents

162884

Sigma-Aldrich

3,4,7,8-Tetramethyl-1,10-phenanthroline

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16N2
CAS Number:
Molecular Weight:
236.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

reaction suitability

reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

mp

277-280 °C (lit.)

solubility

95% ethanol: soluble 50 mg/mL, clear to hazy, colorless to dark yellow

SMILES string

Cc1cnc2c(ccc3c(C)c(C)cnc23)c1C

InChI

1S/C16H16N2/c1-9-7-17-15-13(11(9)3)5-6-14-12(4)10(2)8-18-16(14)15/h5-8H,1-4H3

InChI key

NPAXPTHCUCUHPT-UHFFFAOYSA-N

General description

3,4,7,8-Tetramethyl-1,10-phenanthroline is a metal-chelating agent. It acts as ligand and forms dinuclear Cu(II) hypocrellin B complexes[1]. It also forms tetraaqua(3,4,7,8-tetramethyl-1,10-phenanthroline-kappa2N,N′)zinc(II) thiosulfate complex with zinc[2].

Application

3,4,7,8-Tetramethyl-1,10-phenanthroline was used in the synthesis of heteroleptic cationic Ir(III) complex, 3,4,7,8-tetramethyl-1,10-phenanthroline-bis[2-(2′,4′-difluorophenyl)pyridine]iridium(III) hexafluorophosphate[3].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Actions of an inert nickel (II) chelate of tetramethyl-1,10-phenanthroline at calcium-dependent sites on the guinea-pig isolated atrium.
H J Grossman et al.
Methods and findings in experimental and clinical pharmacology, 1(4), 239-260 (1979-10-01)
Origin of the large spectral shift in electroluminescence in a blue light emitting cationic iridium (III) complex.
Bolink HJ, et al.
Journal of Materials Chemistry, 17(48), 5032-5041 (2007)
Yi Sun et al.
Inorganic chemistry, 49(21), 10108-10116 (2010-09-30)
Five new dinuclear Cu(II) complexes were designed and synthesized, using hypocrellin B, a naturally occurring photosensitizer that has received extensive studies as promising photodynamic therapy (PDT) agent, as bridging ligand, and five kinds of diimine ligands, including 2,2'-bipyridine (bpy), 1,10-phenanthroline
Pui-Kei Lee et al.
Dalton transactions (Cambridge, England : 2003), 40(10), 2180-2189 (2010-08-19)
Four new luminescent cyclometallated iridium(III) bis(quinolylbenzaldehyde) diimine complexes [Ir(qba)(2)(N⁁N)](PF(6)) (Hqba = 4-(2-quinolyl)benzaldehyde, N⁁N = 2,2'-bipyridine, bpy (1); 1,10-phenanthroline, phen (2); 3,4,7,8-tetramethyl-1,10-phenanthroline, Me(4)-phen (3); 4,7-diphenyl-1,10-phenanthroline, Ph(2)-phen (4)) have been synthesised and characterised, and their electronic absorption, emission and electrochemical properties investigated.
M Enriqueta Díaz de Vivar et al.
Acta crystallographica. Section C, Crystal structure communications, 60(Pt 9), m451-m453 (2004-09-04)
In the title complex of zinc(II) with 3,4,7,8-tetramethyl-1,10-phenanthroline (tmph), viz. [Zn(C16H16N2)(H2O)4](S2O3), the metal atom has a monomeric octahedral ZnN2O4 complex environment comprising two N-atom donors from the tmph group and four aqua O-atom donors. The complex cation is connected to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service