Skip to Content
Merck
All Photos(2)

Key Documents

160954

Sigma-Aldrich

Methyl 4-nitrobenzenesulfonate

99%

Synonym(s):

Methyl nosylate, Methyl p-nitrobenzenesulfonate, Methyl p-nitrotosylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€118.00
10 G
€457.00

€118.00


Estimated to ship on05 April 2025


Request a Bulk Order

Select a Size

Change View
1 G
€118.00
10 G
€457.00

About This Item

Linear Formula:
O2NC6H4SO3CH3
CAS Number:
Molecular Weight:
217.20
Beilstein:
2277327
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€118.00


Estimated to ship on05 April 2025


Request a Bulk Order

Assay

99%

form

solid

mp

89-92 °C (lit.)

solubility

acetone: soluble 5%, clear, faintly yellow to greenish-yellow

functional group

nitro
sulfonic acid

SMILES string

COS(=O)(=O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C7H7NO5S/c1-13-14(11,12)7-4-2-6(3-5-7)8(9)10/h2-5H,1H3

InChI key

RMNJNEUWTBBZPT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Reaction between methyl 4-nitrobenzenesulfonate and bromide ions has been studied in mixed single-chain-gemini micellar solutions.[1] Kinetics of SN2 reactions of methyl 4-nitrobenzenesulfonate with ammonia, primary amines, secondary amines, tertiary amines and anionic nucleophiles has been studied.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Lewendon et al.
The Biochemical journal, 290 ( Pt 1), 15-19 (1993-02-15)
A catalytically essential histidine residue (His-195) of chloramphenicol acetyltransferase (CAT) acts as a general base in catalysis, abstracting a proton from the primary hydroxy group of chloramphenicol. The pKa of His-195 has been determined from the pH-dependence of chemical modification.
S Ishii et al.
Protein science : a publication of the Protein Society, 7(8), 1802-1810 (1999-03-19)
Aromatic L-amino acid decarboxylase (AADC) catalytic mechanism has been proposed to proceed through two consecutive intermediates (i.e., Michaelis complex and the external aldimine). Limited proteolysis of AADC that preferentially digested at the C-terminal side of Arg334 was slightly retarded in
O Paquatte et al.
Photochemistry and photobiology, 50(6), 817-825 (1989-12-01)
Vibrio harveyi luciferase, an alpha beta dimer, was effectively inactivated by treatment with the methylation agent methyl p-nitrobenzene sulfonate. However, inactivation of luciferase in the presence of excess amounts of this reagent did not follow pseudo-first-order kinetics. After taking the
R P Swenson et al.
The Journal of biological chemistry, 259(9), 5585-5590 (1984-05-10)
Incubation of D-amino acid oxidase with excess methyl-p-nitrobenzenesulfonate results in a pseudo-first order, irreversible loss of 95% of the assayable activity using D-phenylglycine as substrate. The rate of inactivation reaches a limiting value of 0.021 min-1 (pH 7.7, 22 degrees
María del Mar Graciani et al.
Journal of colloid and interface science, 328(2), 324-330 (2008-10-09)
The reaction between methyl 4-nitrobenzenesulfonate and bromide ions has been studied in mixed single-chain-gemini micellar solutions of n-dodecyltrimethylammonium bromide, DTAB, and dodecyl tricosaoxyethylene glycol ether, Brij(35), with alkanediyl-alpha-omega-bis(dodecyldimethylammonium) bromide, 12-s-12,2Br(-) (s=3,4,5). Kinetic micellar effects show that an increase in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service