Skip to Content
Merck
All Photos(1)

Key Documents

159735

Sigma-Aldrich

N,N′,N′′-Methylidynetrisformamide

97%

Synonym(s):

Tris-formaminomethane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€216.00

€216.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€216.00

About This Item

Linear Formula:
(HCONH)3CH
CAS Number:
Molecular Weight:
145.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€216.00


Estimated to ship on21 May 2025


Request a Bulk Order

Assay

97%

form

solid

mp

170-172 °C (dec.) (lit.)

solubility

water: soluble 25 mg/mL, clear, colorless

functional group

amide

SMILES string

[H]C(=O)NC(NC([H])=O)NC([H])=O

InChI

1S/C4H7N3O3/c8-1-5-4(6-2-9)7-3-10/h1-4H,(H,5,8)(H,6,9)(H,7,10)

InChI key

HNPCDFJZADHNHD-UHFFFAOYSA-N

General description

N,N′,N′′-Methylidynetrisformamide is also referred as tris(formamido)methane.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of Tris (carboalkoxyamino) methane and N-Carbethoxyiminocarboxylic Acid Esters.
Grakauskas V.
The Journal of Organic Chemistry, 36(21), 3251-3252 (1971)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service