Skip to Content
Merck
All Photos(1)

Key Documents

15554

Sigma-Aldrich

Boc-Cys(Npys)-OH

≥99.0% (TLC)

Synonym(s):

Nα-Boc-S-(3-nitro-2-pyridylthio)-L-cysteine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N3O6S2
CAS Number:
Molecular Weight:
375.42
Beilstein:
3569570
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.0% (TLC)

optical activity

[α]20/D −94±5°, c = 1% in methanol

mp

~160 °C (dec.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CSSc1ncccc1[N+]([O-])=O)C(O)=O

InChI

1S/C13H17N3O6S2/c1-13(2,3)22-12(19)15-8(11(17)18)7-23-24-10-9(16(20)21)5-4-6-14-10/h4-6,8H,7H2,1-3H3,(H,15,19)(H,17,18)/t8-/m0/s1

InChI key

OVTLOLNDKQUMRH-QMMMGPOBSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Mourier et al.
Journal of immunological methods, 171(1), 65-71 (1994-05-02)
Some T cell epitopes become inactive when their thiols are blocked with various irreversible reagents (Régnier-Vigouroux, 1988; Maillère, 1992; Maillère et al., 1993). Blocking protein and peptide thiols with BCNP (Boc-Cys(Npys)-OH) constitutes a most appropriate strategy when searching for thiol-containing
M S Bernatowicz et al.
International journal of peptide and protein research, 28(2), 107-112 (1986-08-01)
The 3-nitro-2-pyridinesulfenyl (Npys) derivative of cysteine was prepared and used to facilitate the formation of an unsymmetrical disulfide bond. Since this derivative is stable in trifluoroacetic acid:CH2 Cl2 (1:1) and anhydrous hydrogen fluoride, Boc-Cys(Npys) could be used directly in solid
J W Drijfhout et al.
International journal of peptide and protein research, 32(3), 161-166 (1988-09-01)
The disulfide bond in S-3-nitro-2-pyridinesulfenyl (S-Npys) compounds is stable towards the acid treatment used in solid-phase peptide synthesis, yet the liability of S-Npys-peptides towards nucleophiles enables the conjugation to proteins to proceed under mild conditions. Thus Boc-Cys(Npys)-OH was coupled as
Y Kitamura et al.
Molecular pharmacology, 38(2), 184-191 (1990-08-01)
To gain insight into the coupling mechanism of inhibitory receptors, 5-hydroxytryptamine1A receptors and alpha 2-adrenoceptors, with GTP-binding proteins (G proteins) in the central nervous system, we examined the effects of two 3-nitro-2-pyridinesulfenyl compounds, S-(3-nitro-2-pyridinesulfenyl)-L-cysteine [Cys(Npys)] and N-t-butoxy-carbonyl-S-(3-nitro-2-pyridinesulfenyl)-L-cysteine [Boc-Cys(Npys)], on 1)
Ayami Matsushima et al.
Biopolymers, 106(4), 460-469 (2016-06-09)
We previously showed that an antagonist-based peptide ligand, H-Cys(Npys)-Arg-Tyr-Tyr-Arg- Ile-Lys-NH2 , captures the free thiol groups in the ligand-binding site of the nociceptin receptor ORL1. However, the exact receptor sites of this thiol-disulfide exchange reaction have not been uncovered, although

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service