Skip to Content
Merck
All Photos(1)

Key Documents

155136

Sigma-Aldrich

Chlorocyclopentane

99%

Synonym(s):

Cyclopentyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€139.00

€139.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€139.00

About This Item

Empirical Formula (Hill Notation):
C5H9Cl
CAS Number:
Molecular Weight:
104.58
Beilstein:
1900557
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€139.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.4512 (lit.)

bp

114 °C (lit.)

density

1.005 g/mL at 25 °C (lit.)

SMILES string

ClC1CCCC1

InChI

1S/C5H9Cl/c6-5-3-1-2-4-5/h5H,1-4H2

InChI key

NDTCXABJQNJPCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chlorocyclopentane was used in the preparation of diastereomeric activation products[1].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew J Vetter et al.
Journal of the American Chemical Society, 131(30), 10742-10752 (2009-04-03)
Several halogenated substrates are found to participate in C-H bond cleavage reactions with the photochemically generated fragment [Tp'Rh(CNR)] (Tp' = hydrotris(3,5-dimethylpyrazolyl)borate; R = CH(2)CMe(3)). Reaction with 1- or 3-chloropentane gives only terminal C-H activation products. Reaction with 2-chloropentane gives a
Laura A Mertens et al.
The journal of physical chemistry. A, 122(24), 5418-5436 (2018-05-09)
The relative rates of C-C and C-H β-scission reactions of isobutyl radicals (2-methylprop-1-yl, C4H9) were investigated with shock tube experiments at temperatures of (950 to 1250) K and pressures of (200 to 400) kPa. We produced isobutyl radicals from the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service