Skip to Content
Merck
All Photos(3)

Documents

471542

Sigma-Aldrich

Ethanesulfonyl chloride

≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2SO2Cl
CAS Number:
Molecular Weight:
128.58
Beilstein:
773865
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

refractive index

n20/D 1.452 (lit.)

bp

177 °C (lit.)

density

1.357 g/mL at 25 °C (lit.)

SMILES string

CCS(Cl)(=O)=O

InChI

1S/C2H5ClO2S/c1-2-6(3,4)5/h2H2,1H3

InChI key

FRYHCSODNHYDPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service