Skip to Content
Merck
All Photos(2)

Key Documents

143626

Sigma-Aldrich

m-Tolyl isocyanate

99%

Synonym(s):

3-Methylphenyl isocyanate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€133.00

€133.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€133.00

About This Item

Linear Formula:
CH3C6H4NCO
CAS Number:
Molecular Weight:
133.15
Beilstein:
386149
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€133.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.5305 (lit.)

bp

75-76 °C/12 mmHg (lit.)

density

1.033 g/mL at 25 °C (lit.)

functional group

isocyanate

storage temp.

2-8°C

SMILES string

Cc1cccc(c1)N=C=O

InChI

1S/C8H7NO/c1-7-3-2-4-8(5-7)9-6-10/h2-5H,1H3

InChI key

CPPGZWWUPFWALU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

m-Tolyl isocyanate undergoes polymerization reaction to form the corresponding polymers by using lanthanum isopropoxide as initiator[1].

Application

m-Tolyl isocyanate was used as reagent during the synthesis of C5-cyclohexyl analog of the cholecystokinin type-B receptor antagonist L-365,260[2].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1C - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L-708,474: The C5-cyclohexyl analogue of L-365,260, a selective high affinity ligand for the CCKB/gastrin receptor.
Chambers MS, et al.
Bioorganic & Medicinal Chemistry Letters, 3(10), 1919-1924 (1993)
Lanthanoid isopropoxide as a novel initiator for anionic polymerization of isocyanates.
Fukuwatari N, et al.
Macromolecular Rapid Communications, 17(1), 1-7 (1996)
Lili Zhang et al.
Journal of chromatography. A, 1623, 461174-461174 (2020-06-09)
A new class of chitosan derivatives with an isopropylthiourea at the 2-position and various carbamates at the 3,6-positions of the glucosamine skeleton was synthesized by the selective thiocarbamoylation of the 2-amino group. The chiral stationary phases (CSPs) were then prepared

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service