Skip to Content
Merck
All Photos(2)

Key Documents

131873

Sigma-Aldrich

Tetrachlorothiophene

99%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€123.00

€123.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€123.00

About This Item

Empirical Formula (Hill Notation):
C4Cl4S
CAS Number:
Molecular Weight:
221.92
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€123.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

refractive index

n20/D 1.591 (lit.)

bp

75 °C/2 mmHg (lit.)

mp

28-30 °C (lit.)

density

1.704 g/mL at 25 °C (lit.)

functional group

chloro

SMILES string

Clc1sc(Cl)c(Cl)c1Cl

InChI

1S/C4Cl4S/c5-1-2(6)4(8)9-3(1)7

InChI key

WZXXZHONLFRKGG-UHFFFAOYSA-N

Application

Solvent for organic reactions and spectral use.
Tetrachlorothiophene was used in the preparation of 3,4,5-trichloro-2-thiophenecarbonyl chloride[1].

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John W Hull et al.
Beilstein journal of organic chemistry, 3, 23-23 (2007-09-06)
Dow AgroSciences has been investigating a new family of functionalized 2,6-dihaloaryl 1,2,4-triazole insecticides featuring specifically targeted insecticidal activities coupled with low mammalian toxicity. With broad spectrum control of both chewing and sap-feeding pests in mind, this family of compounds has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service