Skip to Content
Merck
All Photos(1)

Key Documents

125210

Sigma-Aldrich

3-Chloroquinuclidine hydrochloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12ClN · HCl
CAS Number:
Molecular Weight:
182.09
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

236-239 °C (lit.)

functional group

chloro

SMILES string

Cl.ClC1CN2CCC1CC2

InChI

1S/C7H12ClN.ClH/c8-7-5-9-3-1-6(7)2-4-9;/h6-7H,1-5H2;1H

InChI key

OOMINTVSHDCDRK-UHFFFAOYSA-N

Application

3-Chloroquinuclidine hydrochloride has been used in determination of equilibrium constants in D2O for the formation of imines/iminium ions from addition of glycine methyl ester to acetone and from addition of glycine to phenylglyoxylate by 1H-NMR analysis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Crugeiras et al.
Journal of the American Chemical Society, 130(6), 2041-2050 (2008-01-18)
Equilibrium constants in D2O were determined by 1H NMR analyses for formation of imines/iminium ions from addition of glycine methyl ester to acetone and from addition of glycine to phenylglyoxylate. First-order rate constants, also determined by 1H NMR, are reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service