Skip to Content
Merck
All Photos(1)

Key Documents

122394

Sigma-Aldrich

Thiochroman-4-one

97%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
€200.00

€200.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
€200.00

About This Item

Empirical Formula (Hill Notation):
C9H8OS
CAS Number:
Molecular Weight:
164.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€200.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

refractive index

n20/D 1.6395 (lit.)

bp

154 °C/12 mmHg (lit.)

mp

28-30 °C (lit.)

functional group

ketone
thioether

SMILES string

O=C1CCSc2ccccc12

InChI

1S/C9H8OS/c10-8-5-6-11-9-4-2-1-3-7(8)9/h1-4H,5-6H2

InChI key

CVQSWZMJOGOPAV-UHFFFAOYSA-N

Application

Thiochroman-4-one was used as the starting reactant for synthesis of series of pyrazoles and isoxazoles[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Ramalingam et al.
Journal of medicinal chemistry, 20(6), 847-850 (1977-06-01)
A small series of pyrazoles and isoxazoles derived from thiochroman-4-one has been synthesized and characterized. The compounds were examined for their in vitro inhibitory activity against Bacillus subtilis and Pseudomonas fluorescens. Among the tested compounds the pyrazole derivative from thiochroman-4-one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service