Skip to Content
Merck
All Photos(1)

Key Documents

120693

Sigma-Aldrich

α-Methyl-DL-tyrosine

98%

Synonym(s):

DL-2-Methyl-3-(4-hydroxyphenyl)alanine, AMPT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(HO)C6H4CH2C(CH3)(NH2)CO2H
CAS Number:
Molecular Weight:
195.22
Beilstein:
2938704
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

98%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

technique(s)

HPLC: suitable

color

white to off-white

mp

>300 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(N)(Cc1ccc(O)cc1)C(O)=O

InChI

1S/C10H13NO3/c1-10(11,9(13)14)6-7-2-4-8(12)5-3-7/h2-5,12H,6,11H2,1H3,(H,13,14)

InChI key

NHTGHBARYWONDQ-UHFFFAOYSA-N

Gene Information

human ... TH(7054)

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khoa D Nguyen et al.
Nature, 480(7375), 104-108 (2011-11-22)
All homeotherms use thermogenesis to maintain their core body temperature, ensuring that cellular functions and physiological processes can continue in cold environments. In the prevailing model of thermogenesis, when the hypothalamus senses cold temperatures it triggers sympathetic discharge, resulting in
Joram D Mul et al.
PloS one, 8(3), e60214-e60214 (2013-04-05)
The orexigenic neuropeptide melanin-concentrating hormone (MCH), a product of Pmch, is an important mediator of energy homeostasis. Pmch-deficient rodents are lean and smaller, characterized by lower food intake, body-, and fat mass. Pmch is expressed in hypothalamic neurons that ultimately
Maria Xilouri et al.
PloS one, 4(5), e5515-e5515 (2009-05-14)
The mechanisms through which aberrant alpha-synuclein (ASYN) leads to neuronal death in Parkinson's disease (PD) are uncertain. In isolated liver lysosomes, mutant ASYNs impair Chaperone Mediated Autophagy (CMA), a targeted lysosomal degradation pathway; however, whether this occurs in a cellular
Sunhoe Bang et al.
PLoS genetics, 7(3), e1001346-e1001346 (2011-04-02)
The ability to respond to environmental temperature variation is essential for survival in animals. Flies show robust temperature-preference behaviour (TPB) to find optimal temperatures. Recently, we have shown that Drosophila mushroom body (MB) functions as a center controlling TPB. However
Kinuyo Iwata et al.
The Journal of reproduction and development, 57(3), 379-384 (2011-03-02)
Ketosis is found in various pathophysiological conditions, including diabetes and starvation, that are accompanied by suppression of gonadal activity. The aim of the present study was to determine the role of ketone body in the brain in regulating pulsatile luteinizing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service