Skip to Content
Merck
All Photos(1)

Key Documents

118613

Sigma-Aldrich

2-Methyl-2-heptene

98%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€119.00

€119.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€119.00

About This Item

Linear Formula:
CH3(CH2)3CH=C(CH3)2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1732139
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€119.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

refractive index

n20/D 1.417 (lit.)

bp

122 °C (lit.)

mp

−122 °C (lit.)

density

0.724 g/mL at 25 °C (lit.)

SMILES string

CCCC\C=C(\C)C

InChI

1S/C8H16/c1-4-5-6-7-8(2)3/h7H,4-6H2,1-3H3

InChI key

WEPNJTDVIIKRIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-2-heptene undergoes oxidation by singlet oxygen generated by the excitation of thiazine dyes cation to form hydroperoxides[1].

Application

2-Methyl-2-heptene was used in the functionalization of ethylene/5,7-dimethyl-1,6-diene copolymer with maleic anhydride via Alder Ene reaction[2].

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Singlet oxygen mediated oxidation of olefins within zeolites: selectivity and complexities.
Shailaja J, et al.
Tetrahedron, 56(36), 6927-6943 (2000)
1H-NMR study of maleic anhydride modified ethylene-diene copolymers.
Vicente AI, et al.
Journal of Polymer Research, 18(4), 527-532 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service