Skip to Content
Merck
All Photos(1)

Key Documents

111058

Sigma-Aldrich

2-Methyl-1-heptene

97%

Synonym(s):

2-Methyl-1-heptene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€840.00

€840.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€840.00

About This Item

Linear Formula:
CH3(CH2)4C(CH3)=CH2
CAS Number:
Molecular Weight:
112.21
Beilstein:
1733419
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€840.00


Check Cart for Availability

Request a Bulk Order

vapor pressure

38 mmHg ( 37.7 °C)

Quality Level

Assay

97%

refractive index

n20/D 1.411 (lit.)

bp

117-120 °C (lit.)

density

0.713 g/mL at 25 °C (lit.)

SMILES string

CCCCCC(C)=C

InChI

1S/C8H16/c1-4-5-6-7-8(2)3/h2,4-7H2,1,3H3

InChI key

RCBGGJURENJHKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Methyl-1-heptene was used to study the epoxidation of olefins with H2O2; where ionic liquid ( e.g. 1-butyl-3-methylimidazolium hexafluorophosphate) was an activator and Keggin polyoxometalate [bmim]3PW12O40 was the catalyst[1].

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

50.0 °F - closed cup

Flash Point(C)

10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liu L, et al.
New. J. Chem., 32(2), 283-289 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service