Skip to Content
Merck
All Photos(1)

Documents

114685

Sigma-Aldrich

4-Nitrophenylhydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4NHNH2
CAS Number:
Molecular Weight:
153.14
Beilstein:
608107
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

solid

contains

≥10% water as stabilizer

mp

156 °C (dec.) (lit.)

SMILES string

NNc1ccc(cc1)[N+]([O-])=O

InChI

1S/C6H7N3O2/c7-8-5-1-3-6(4-2-5)9(10)11/h1-4,8H,7H2

InChI key

KMVPXBDOWDXXEN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exploding BombExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Expl. 1.1 - Eye Irrit. 2 - Flam. Sol. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

1 - Explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R V Raju et al.
Journal of AOAC International, 77(3), 748-751 (1994-05-01)
Three spectrophotometric methods were developed for the microdetermination of decamethrin in insecticidal formulations and in water. The methods are based on the hydrolysis of decamethrin with methanolic KOH to 3-phenoxybenzaldehyde; condensation of the hydrolysis product with 2,4-dinitrophenylhydrazine (2,4-DNPH), 4-nitrophenyl-hydrazine (4-NPH)
Minae Mure et al.
Journal of the American Chemical Society, 125(20), 6113-6125 (2003-06-06)
4-n-Butylamino-5-ethyl-1,2-benzoquinone (1(ox)) has been synthesized as a model compound for the LTQ (lysine tyrosyl quinone) cofactor of lysyl oxidase (LOX). At pH 7, 1(ox) has a lambda(max) at 504 nm and exists as a neutral o-quinone in contrast to a
N Nakamura et al.
The Journal of biological chemistry, 271(9), 4718-4724 (1996-03-01)
Resonance Raman spectroscopy is an excellent technique for providing structural information on the 2,4, 5-trihydroxyphenylalanine quinone (TPQ) cofactor in copper-containing amine oxidases. This technique has been used to investigate the copper- and O2-dependent biosynthesis of the TPQ cofactor in phenylethylamine
T Huque et al.
Comparative biochemistry and physiology. B, Comparative biochemistry, 86(1), 135-139 (1987-01-01)
The total plasmalogen content of lingual and other tissues was analyzed using the iodine-addition (Method 1), the p-nitrophenylhydrazone (Method 2), and the two-dimensional thin layer chromatography procedure (Method 3). Methods 1 and 2 were simple, rapid and reproducible, yielding values
Y Kariya et al.
Journal of biochemistry, 123(2), 240-246 (1998-04-16)
Compositional analyses of heparin (Hep) and heparan sulfate (HS) have been undertaken with disaccharide units obtained by either enzymatic digestion with heparitinases or hydrazinolysis/deamination reaction of polysaccharides. Unsaturated disaccharide units generated by the enzymatic method are detectable on HPLC with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service