Skip to Content
Merck
All Photos(1)

Documents

113220

Sigma-Aldrich

1-Phenylundecane

99%

Synonym(s):

Undecylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)10CH3
CAS Number:
Molecular Weight:
232.40
Beilstein:
2044467
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.482 (lit.)

bp

316 °C (lit.)

mp

−5 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCCCc1ccccc1

InChI

1S/C17H28/c1-2-3-4-5-6-7-8-9-11-14-17-15-12-10-13-16-17/h10,12-13,15-16H,2-9,11,14H2,1H3

InChI key

XBEADGFTLHRJRB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Undecylbenzene (1-Phenylundecane) forms a charge-transfer complex with fluoranil.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

K Elias Ikonen et al.
Journal of chromatography. A, 1614, 460705-460705 (2019-11-23)
An update for ASTM D2425, "Standard Test Method for Hydrocarbon Types in Middle Distillates by Mass Spectrometry," was recently released to expand its scope by adding modern mass spectrometric instrumentation and a new sample type, synthesized hydrocarbons. While the method

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service