Skip to Content
Merck
All Photos(2)

Key Documents

112283

Sigma-Aldrich

Ethyl isovalerate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€30.90

€30.90


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
100 ML
€30.90

About This Item

Linear Formula:
(CH3)2CHCH2COOC2H5
CAS Number:
Molecular Weight:
130.18
Beilstein:
1744677
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39022323
PubChem Substance ID:
NACRES:
NA.22

€30.90


Estimated to ship on01 April 2025


Request a Bulk Order

vapor pressure

7.5 mmHg ( 20 °C)

Quality Level

Assay

98%

refractive index

n20/D 1.396 (lit.)

bp

131-133 °C (lit.)

mp

−99 °C (lit.)

density

0.864 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)CC(C)C

InChI

1S/C7H14O2/c1-4-9-7(8)5-6(2)3/h6H,4-5H2,1-3H3

InChI key

PPXUHEORWJQRHJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vrushali Dandavate et al.
Journal of microbiology and biotechnology, 18(4), 735-741 (2008-05-10)
In our previous study, a surfactant-coated Candida rugosa lipase immobilized in microemulsion-based organogels was exploited for the synthesis of ethyl isovalerate. In the present study, we are focusing on the effective reuse of lipase immobilized in microemulsion-based organogels (MBGs) in
Yong-Hyun Kim et al.
Sensors (Basel, Switzerland), 13(6), 7939-7978 (2013-06-22)
The classes and concentrations of volatile organic compounds (VOC) released from fresh and decaying strawberries were investigated and compared. In this study, a total of 147 strawberry volatiles were quantified before and after nine days of storage to explore differences
Johannes Niebler et al.
Phytochemistry, 109, 66-75 (2014-12-04)
Frankincense has been known, traded and used throughout the ages for its exceptional aroma properties, and is still commonly used in both secular and religious settings to convey a pleasant odor. Surprisingly, the odoriferous principle(s) underlying its unique odor profile
Amélie Slegers et al.
Molecules (Basel, Switzerland), 20(6), 10980-11016 (2015-06-18)
Developed from crosses between Vitis vinifera and North American Vitis species, interspecific hybrid grape varieties are becoming economically significant in northern areas, where they are now extensively grown for wine production. However, the varietal differences between interspecific hybrids are not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service