Skip to Content
Merck
All Photos(1)

Key Documents

110671

Sigma-Aldrich

2,2-Dimethylpentane

≥99%

Synonym(s):

1,1,1-Trimethylbutane

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€159.00
10 G
€302.00

€159.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€159.00
10 G
€302.00

About This Item

Linear Formula:
CH3CH2CH2C(CH3)3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730757
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€159.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

expl. lim.

~8.3 %

refractive index

n20/D 1.382 (lit.)

bp

78 °C/743 mmHg (lit.)

density

0.674 g/mL at 25 °C (lit.)

SMILES string

CCCC(C)(C)C

InChI

1S/C7H16/c1-5-6-7(2,3)4/h5-6H2,1-4H3

InChI key

CXOWYJMDMMMMJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,2-Dimethylpentane is a component of the corona discharge of a simulated Titan′s atmosphere[1].

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Navarro-Gonzalez et al.
Uchu Seibutsu Kagaku, 12(2), 81-91 (2001-09-07)
The atmosphere of Titan is constantly bombarded by galactic cosmic rays and Saturnian magnetospheric electrons causing the formation of free electrons and primary ions, which are then stabilized by ion cluster formation and charging of aerosols. These charged particles accumulate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service