Skip to Content
Merck
All Photos(1)

Key Documents

11010

Sigma-Aldrich

Aluminum

powder (fine), ≥93% (complexometric)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Al
CAS Number:
Molecular Weight:
26.98
EC Number:
MDL number:
UNSPSC Code:
12141702
PubChem Substance ID:

Assay

≥93% (complexometric)

form

powder (fine)

autoignition temp.

1400 °F

reaction suitability

reagent type: catalyst
core: aluminum

resistivity

2.6548 μΩ-cm

impurities

≤0.03% heavy metals (as Pb)
≤0.3% soluble in ether

bp

2460 °C (lit.)

mp

660.37 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

cation traces

Fe: ≤5000 mg/kg

SMILES string

[Al]

InChI

1S/Al

InChI key

XAGFODPZIPBFFR-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Water-react 2

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

Structural features of oxide coatings on aluminum.
Keller F, et al.
Journal of the Electrochemical Society, 100(9), 411-419 (1953)
Electrochemistry of Cd (II) in the basic 1-ethyl-3-methylimidazolium chloride/tetrafluoroborate room temperature molten salt.
Chen P-Y and Sun I-W
Electrochimica Acta, 45(19), 3163-3170 (2000)
Aluminum ion diffusion in aluminum oxide.
Paladino AE and Kingery WD.
J. Chem. Phys. , 37(5), 957-962 (1962)
Carlos Eduardo Flores-Tinoco et al.
Molecular systems biology, 16(6), e9419-e9419 (2020-06-04)
Biological nitrogen fixation emerging from the symbiosis between bacteria and crop plants holds promise to increase the sustainability of agriculture. One of the biggest hurdles for the engineering of nitrogen-fixing organisms is an incomplete knowledge of metabolic interactions between microbe
Thierry Deschamps et al.
Optics express, 21(7), 8382-8392 (2013-04-11)
Using a combination of experimental techniques such as optical absorption, Raman scattering, continuous wave and pulse Electron Spin Resonance (ESR), we characterize a set of γ-irradiated Yb(3+) doped silica glass preforms with different contents of phosphorous and aluminum. We demonstrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service