Skip to Content
Merck
All Photos(1)

Key Documents

109371

Sigma-Aldrich

Bicyclo[2.2.1]hept-5-ene-2-carboxaldehyde

95%

Synonym(s):

5-Norbornene-2-carboxaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€138.00

€138.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€138.00

About This Item

Empirical Formula (Hill Notation):
C8H10O
CAS Number:
Molecular Weight:
122.16
Beilstein:
1363813
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€138.00


Check Cart for Availability

Request a Bulk Order

Assay

95%

refractive index

n20/D 1.490 (lit.)

bp

67-70 °C/12 mmHg (lit.)

density

1.03 g/mL at 20 °C (lit.)

SMILES string

O=CC1C[C@H]2C[C@@H]1C=C2

InChI

1S/C8H10O/c9-5-8-4-6-1-2-7(8)3-6/h1-2,5-8H,3-4H2/t6-,7+,8?/m1/s1

InChI key

AJIBZRIAUXVGQJ-KVARREAHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5-Norbornene-2-carboxaldehyde was used in transition metal-catalyzed synthesis of norbornene-derived homopolymers having pendent t-Boc-protected quinizarin moieties for patterned fluorescence images[1].

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The first synthesis of a transition metal-catalyzed homopolymer having pendentt-boc-protected quinizarin for patterned fluorescence images.
Yoo J, et al.
Macromolecular Research, 11(1), 69-72 (2003)
Cheng Zhang et al.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry, 21(8), 1047-1060 (2016-11-01)
A new ligand BTCP and its iridium(III) complex [Ir(ppy)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service