Skip to Content
Merck
All Photos(2)

Key Documents

108235

Sigma-Aldrich

2-tert-Butylanthraquinone

98%

Synonym(s):

β- tert -Butylanthraquinone, 2-(1,1-Dimethylethyl)-9,10-anthracenedione, 2-tert -Butyl-9,10-anthraquinone

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€238.00

€238.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€238.00

About This Item

Empirical Formula (Hill Notation):
C18H16O2
CAS Number:
Molecular Weight:
264.32
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

€238.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

form

solid

mp

98-100 °C (lit.)

SMILES string

CC(C)(C)c1ccc2C(=O)c3ccccc3C(=O)c2c1

InChI

1S/C18H16O2/c1-18(2,3)11-8-9-14-15(10-11)17(20)13-7-5-4-6-12(13)16(14)19/h4-10H,1-3H3

InChI key

YTPSFXZMJKMUJE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymerization of Methyl Methacrylate Photoinitiated by Anthraquinone and 2-tert-Butylanthraquinone
Ledwith, A., G. Ndaalio, and A. R. Taylor
Macromolecules, 8(1), 1-7 (1975)
Liquid chromatographic detection of cardiac glycosides, saccharides and hydrocortisone based on the photoreduction of 2-tert-butylanthraquinone
Gandelman, M. S.
Analytica Chimica Acta, 155, 159-171 (1983)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service