Skip to Content
Merck
All Photos(2)

Key Documents

180203

Sigma-Aldrich

Borane morpholine complex

95%

Synonym(s):

Morpholineborane, NSC 93813, Trihydro(morpholine-N4)boron

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€46.80

€46.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€46.80

About This Item

Empirical Formula (Hill Notation):
C4H12BNO
CAS Number:
Molecular Weight:
100.96
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

€46.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

reaction suitability

reagent type: reductant

mp

97-99 °C (lit.)

functional group

ether

SMILES string

B.C1COCCN1

InChI

1S/C4H9NO.BH3/c1-3-6-4-2-5-1;/h5H,1-4H2;1H3

InChI key

IFVAHVOGOBRFSP-UHFFFAOYSA-N

Application

Reactant involved in:
  • Ethanolysis producing a reducing system
  • Being a reducing agent for synthesis of Rh nanoparticle / carbon nanotube catalysts
  • Stereoselective and regioselective reduction of steroidal ketones
  • Methanolic cleavage

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Anna L Brown et al.
Nanoscale research letters, 13(1), 208-208 (2018-07-15)
High-density inorganic nanoparticles have shown promise in medical applications that utilize radiation including X-ray imaging and as radiation dose enhancers for radiotherapy. We have developed an aqueous synthetic method to produce small (~ 2 nm) iridium nanoparticles (IrNPs) by reduction of iridium(III)
Sibel Eken Korkut et al.
ACS applied materials & interfaces, 12(7), 8130-8139 (2020-01-28)
A novel ternary nanocomposite, mesoporous graphitic carbon nitride/black phosphorus-AgPd (denoted mpg-CN/BP-AgPd), was successfully fabricated by assembling the as-prepared AgPd alloy nanoparticles (NPs) on mesoporous graphitic carbon nitride/black phosphorus (mpg-CN/BP) binary composites. This novel nanocomposite comprises a heterojunction support material formed
Guangming Jiang et al.
ACS nano, 9(11), 11014-11022 (2015-10-06)
We report the synthesis of core/shell face-centered tetragonal (fct)-FePd/Pd nanoparticles (NPs) via reductive annealing of core/shell Pd/Fe3O4 NPs followed by temperature-controlled Fe etching in acetic acid. Among three different kinds of core/shell FePd/Pd NPs studied (FePd core at ∼8 nm

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service