Skip to Content
Merck
All Photos(1)

Key Documents

104566

Supelco

Thorin

indicator grade

Synonym(s):

1-(2-Arsonophenylazo)-2-naphthol-3,6-disulfonic acid disodium salt, 2-(2-Hydroxy-3,6-disulfo-1-naphthylazo)phenylarsonic acid disodium salt, 2-(3,6-Disulfo-2-hydroxy-1-naphthylazo)benzenearsonic acid disodium salt, APANS, Thoron(ol)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11AsN2Na2O10S2
CAS Number:
Molecular Weight:
576.30
Beilstein:
2957648
EC Number:
MDL number:
UNSPSC Code:
12161500
PubChem Substance ID:

grade

indicator grade

technique(s)

titration: suitable

λmax

483 nm

SMILES string

[Na+].[Na+].Oc1c(\N=N\c2ccccc2[As](O)(O)=O)c3ccc(cc3cc1S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C16H13AsN2O10S2.2Na/c20-16-14(31(27,28)29)8-9-7-10(30(24,25)26)5-6-11(9)15(16)19-18-13-4-2-1-3-12(13)17(21,22)23;;/h1-8,20H,(H2,21,22,23)(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b19-18+;;

InChI key

DCSRPHQBFSYJNN-BUFQOAPZSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service