Skip to Content
Merck
All Photos(1)

Key Documents

100188

Sigma-Aldrich

2,3,5,6-Tetrafluorobenzenethiol

97%

Synonym(s):

2,3,5,6-Tetrafluorothiophenol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€87.00

€87.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€87.00

About This Item

Linear Formula:
HC6F4SH
CAS Number:
Molecular Weight:
182.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€87.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

storage condition

under inert gas

refractive index

n20/D 1.4865 (lit.)

bp

152-153 °C (lit.)

functional group

fluoro

SMILES string

Fc1cc(F)c(F)c(S)c1F

InChI

1S/C6H2F4S/c7-2-1-3(8)5(10)6(11)4(2)9/h1,11H

InChI key

IGOGJHYWSOZGAE-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

118.4 °F - closed cup

Flash Point(C)

48 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Quernin et al.
Journal of chromatography. B, Biomedical sciences and applications, 709(1), 47-56 (1998-07-08)
A specific and highly sensitive method has been developed for the determination of busulfan in plasma by gas chromatography-mass spectrometry using a deuterium-labeled busulfan (busulfan-d8) as internal standard. Plasma containing busulfan and busulfan-d8 were extracted with ethyl acetate and derivatized
L Embree et al.
Cancer chemotherapy and pharmacology, 32(2), 137-142 (1993-01-01)
The development and validation of a gas chromatographic assay method for determination of total and free busulfan concentrations in human plasma for pharmacokinetic studies is reported. 1,6-Bis(methanesulfonyloxy)hexane, the internal standard, and a potential metabolite, 3-hydroxysulfolane, were synthesized. Plasma and plasma
M H Quernin et al.
Journal of chromatography. B, Biomedical sciences and applications, 721(1), 147-152 (1999-02-23)
A high-performance liquid chromatographic (HPLC) method was developed and validated for the determination of busulfan in plasma. Busulfan was extracted in toluene, derivatized by 2,3,5,6-tetrafluorothiophenol to obtain di-TFTP-butane, the derivatization product was then re-extracted in toluene and injected into the
R B Burns et al.
Journal of pharmaceutical and biomedical analysis, 13(9), 1073-1078 (1995-08-01)
A gas-chromatographic assay method was developed and validated for determination of busulfan in human plasma for test dose therapeutic drug monitoring. Busulfan and the internal standard (1,6-bis-(methanesulfonyloxy)hexane) were extracted from plasma samples and derivatized with 2,3,5,6-tetrafluorothiophenol prior to gas chromatographic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service