Skip to Content
Merck
All Photos(1)

Key Documents

539155

Sigma-Aldrich

4-Fluorobenzyl mercaptan

96%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€37.00

€37.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€37.00

About This Item

Linear Formula:
FC6H4CH2SH
CAS Number:
Molecular Weight:
142.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€37.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

refractive index

n20/D 1.545 (lit.)

bp

72-74 °C/15 mmHg (lit.)

density

1.157 g/mL at 25 °C (lit.)

functional group

fluoro
thiol

SMILES string

Fc1ccc(CS)cc1

InChI

1S/C7H7FS/c8-7-3-1-6(5-9)2-4-7/h1-4,9H,5H2

InChI key

RKTRHMNWVZRZJQ-UHFFFAOYSA-N

General description

4-Fluorobenzyl mercaptan undergoes reaction with p-chloranil to afford mainly 2,5-dichloro-3,6-S-disubstituted benzoquinone and 2,6-dichloro-3,5-S-disubstituted benzoquinone.[1]

Application

4-Fluorobenzyl mercaptan may be used in the synthesis of the degradation products of fluorapacin, namely bis(4-fluorobenzyl)disulfide and bis(4-fluorobenzyl)tetrasulfide.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.0 °F - closed cup

Flash Point(C)

76.67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synthesis of new cyclic thioquinone derivatives.
Ibis C and Ozsoy-Gunes Z.
Heteroatom Chem., 21(6), 446-452 (2010)
Yimei Bao et al.
Journal of pharmaceutical and biomedical analysis, 48(3), 664-671 (2008-08-06)
Bis(4-fluorobenzyl)trisulfide, fluorapacin, has been extensively developed as a promising new anticancer drug candidate. Its degradation products were identified and verified by the newly synthesized compounds bis(4-fluorobenzyl)disulfide (A) and bis(4-fluorobenzyl)tetrasulfide (B) which were resulted from the disproportionation of fluorapacin under forced

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service