Skip to Content
Merck
All Photos(1)

Key Documents

794171

Sigma-Aldrich

9-Mesityl-10-methylacridinium tetrafluoroborate

Synonym(s):

9-Mesityl-10-methylacridinium tetrafluoroborate, 10-Methyl-9-(2,4,6-trimethylphenyl)acridinium tetrafluoroborate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€76.50
1 G
€124.00

€76.50


Estimated to ship on12 April 2025


Request a Bulk Order

Select a Size

Change View
250 MG
€76.50
1 G
€124.00

About This Item

Empirical Formula (Hill Notation):
C23H22BF4N
CAS Number:
Molecular Weight:
399.23
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

€76.50


Estimated to ship on12 April 2025


Request a Bulk Order

form

solid

Quality Level

reaction suitability

core: acridinium
reaction type: Photocatalysis
reagent type: catalyst

photocatalyst activation

450 nm

SMILES string

CC(C=C1C)=CC(C)=C1C2=C3C(C=CC=C3)=[N+](C)C4=CC=CC=C42.F[B-](F)(F)F

InChI

1S/C23H22N.BF4/c1-15-13-16(2)22(17(3)14-15)23-18-9-5-7-11-20(18)24(4)21-12-8-6-10-19(21)23;2-1(3,4)5/h5-14H,1-4H3;/q+1;-1

InChI key

KKMWMXQQOIIJIZ-UHFFFAOYSA-N

Application

Catalyst has been reported by Nicewicz and coworkers to mediate myriad transformations through Photoredox Catalysis. The following applications include the anti-Markovnikov hydroamination of alkenes and addition of carboxylic acids to alkenes and the hydrotrifluoromethylation of styrenes using the Langlois reagent (743232).

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Markovnikov and anti-Markovnikov alkene reactivity differences are discussed, highlighting challenges and catalytic advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service