Creatin-Kinase
![Creatin-Kinase application for HPLC](/deepweb/assets/sigmaaldrich/product/images/301/757/e71c3c66-5281-40fb-aca9-8b610b9e75c4/800/e71c3c66-5281-40fb-aca9-8b610b9e75c4.jpg)
CONDITIONS
column
Discovery BIO PolyMA-SCX, 5 cm × 4.6 mm, 5 μm particles (59601-U)
mobile phase
[A] 20 mM sodium phosphate, pH 7.0; [B] 20 mM sodium phosphate, 0.5M NaCl, pH 7.0
flow rate
0.5 mL/min
injection
10 μL
column temp.
25 °C
detector
UV, 280 nm
Beschreibung
Hinweis zur Analyse
Discovery BIO PolyMA-SCX and PolyMA-WAX columns give sharp, efficient peaks for a wide variety of proteins.