Merck
  • RhoC regulates the actin remodeling required for phagosome formation during FcγR-mediated phagocytosis.
RhoC regulates the actin remodeling required for phagosome formation during FcγR-mediated phagocytosis.
Journal of cell science (2017-11-09)
Youhei Egami, Katsuhisa Kawai, Nobukazu Araki
ZUSAMMENFASSUNG
Phagosome formation is a complicated process that requires spatiotemporally regulated actin reorganization. We found that RhoC GTPase is a critical regulator of FcγR-mediated phagocytosis in macrophages. Our live-cell imaging revealed that RhoC, but not RhoA, is recruited to phagocytic cups engulfing IgG-opsonized erythrocytes (IgG-Es). RhoC silencing through RNAi, CRISPR/Cas-mediated RhoC knockout, and the expression of dominant-negative or constitutively active RhoC mutants suppressed the phagocytosis of IgG-Es. Moreover, RhoC-GTP pulldown experiments showed that endogenous RhoC is transiently activated during phagosome formation. Notably, actin-driven pseudopod extension, which is required for the formation of phagocytic cups, was severely impaired in cells expressing the constitutively active mutant RhoC-G14V, which induced abnormal F-actin accumulation underneath the plasma membrane. mDia1 (encoded by
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
IgG aus Humanserum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder