Merck
  • Basic Residues of β-Sheet A Contribute to Heparin Binding and Activation of Vaspin (Serpin A12).
Basic Residues of β-Sheet A Contribute to Heparin Binding and Activation of Vaspin (Serpin A12).
The Journal of biological chemistry (2016-12-13)
David Ulbricht, Kathrin Oertwig, Kristin Arnsburg, Anja Saalbach, Jan Pippel, Norbert Sträter, John T Heiker
ZUSAMMENFASSUNG
Many members of the serine protease inhibitor (serpin) family are activated by glycosaminoglycans (GAGs). Visceral adipose tissue-derived serpin (vaspin), serpin A12 of the serpin family, and its target protease kallikrein 7 (KLK7) are heparin-binding proteins, and inhibition of KLK7 by vaspin is accelerated by heparin. However, the nature of GAG binding to vaspin is not known. Here, we measured vaspin binding of various glycosaminoglycans and low molecular weight heparins by microscale thermophoresis and analyzed acceleration of protease inhibition by these molecules. In addition, basic residues contributing to heparin binding and heparin activation were identified by a selective labeling approach. Together, these data show that vaspin binds heparin with high affinity (K
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fondaparinux Natrium, ≥95% (HPLC)