Merck
Expression and purification of functional human mu opioid receptor from E.coli.
PloS one (2013-02-26)
Yanbin Ma, Jan Kubicek, Jörg Labahn
ZUSAMMENFASSUNG
N-terminally his-tagged human mu opioid receptor, a G protein-coupled receptor was produced in E.coli employing synthetic codon-usage optimized constructs. The receptor was expressed in inclusion bodies and membrane-inserted in different E.coli strains. By optimizing the expression conditions the expression level for the membrane-integrated receptor was raised to 0.3-0.5 mg per liter of culture. Milligram quantities of receptor could be enriched by affinity chromatography from IPTG induced cultures grown at 18°C. By size exclusion chromatography the protein fraction with the fraction of alpha-helical secondary structure expected for a 7-TM receptor was isolated, by CD-spectroscopy an alpha-helical content of ca. 45% was found for protein solubilised in the detergent Fos-12. Receptor in Fos-12 micelles was shown to bind endomorphin-1 with a K(D) of 61 nM. A final yield of 0.17 mg functional protein per liter of culture was obtained.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Spautin-1, ≥98% (HPLC)