Merck
  • Design, synthesis, and evaluation of cyclic amide/imide-bearing hydroxamic acid derivatives as class-selective histone deacetylase (HDAC) inhibitors.
Design, synthesis, and evaluation of cyclic amide/imide-bearing hydroxamic acid derivatives as class-selective histone deacetylase (HDAC) inhibitors.
Bioorganic & medicinal chemistry (2006-08-01)
Chihiro Shinji, Satoko Maeda, Keisuke Imai, Minoru Yoshida, Yuichi Hashimoto, Hiroyuki Miyachi
ZUSAMMENFASSUNG
A series of hydroxamic acid derivatives bearing a cyclic amide/imide group as a linker and/or cap structure, prepared during our structural development studies based on thalidomide, showed class-selective potent histone deacetylase (HDAC)-inhibitory activity. Structure-activity relationship studies indicated that the steric character of the substituent introduced at the cyclic amide/imide nitrogen atom, the presence of the amide/imide carbonyl group, the hydroxamic acid structure, the shape of the linking group, and the distance between the zinc-binding hydroxamic acid group and the cap structure are all important for HDAC-inhibitory activity and class selectivity. A representative compound (30w) showed potent p21 promoter activity, comparable with that of trichostatin A (TSA), and its cytostatic activity against cells of the human prostate cell line LNCaP was more potent than that of the well-known HDAC inhibitor, suberoylanilide hydroxamic acid (SAHA).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
O-(Tetrahydro-2H-pyran-2-yl)hydroxylamin, 96%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.