Merck
  • Efficacy of short novel antimicrobial and anti-inflammatory peptides in a mouse model of methicillin-resistant Staphylococcus aureus (MRSA) skin infection.
Efficacy of short novel antimicrobial and anti-inflammatory peptides in a mouse model of methicillin-resistant Staphylococcus aureus (MRSA) skin infection.
Drug design, development and therapy (2014-11-08)
Mohamed F Mohamed, Mohamed N Seleem
ZUSAMMENFASSUNG
The therapeutic efficacy of two novel short antimicrobial and anti-inflammatory peptides (RR and RRIKA) was evaluated in a mouse model of staphylococcal skin infection. RR (2%) and RRIKA (2%) significantly reduced the bacterial counts and the levels of proinflammatory cytokines, tumor necrosis factor (TNF)-α, and interleukin (IL)-6, in methicillin-resistant Staphylococcus aureus USA 300-0114 skin lesions. Furthermore, the combined therapy of RRIKA (1%) and lysostaphin (0.5%) had significantly higher antistaphylococcal and anti-inflammatory activity compared to monotherapy. This study supports the potential use of these peptides for topical treatment of methicillin-resistant Staphylococcus aureus skin infections.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fusidinsäure
Fusidinsäure für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Fusidinsäure, European Pharmacopoeia (EP) Reference Standard