Merck
  • Total synthesis of 0231B, an inhibitor of 3alpha-hydroxysteroid dehydrogenase produced by Streptomyces sp. HKI0231.
Total synthesis of 0231B, an inhibitor of 3alpha-hydroxysteroid dehydrogenase produced by Streptomyces sp. HKI0231.
Bioscience, biotechnology, and biochemistry (2003-05-02)
Taichi Komoda, Yoshihiko Shinoda, Shin-ichi Nakatsuka
ZUSAMMENFASSUNG
The new inhibitors of 3alpha-hydroxysteroid dehydrogenase, 0231A 1 and 0231B 2, have a unique benz[c,d]indol-3(1H)-one structure in their molecules. In our advanced studies on indole chemistry, we have developed an efficient synthetic method for benz[c,d]indol-3(1H)-one derivatives. We report here its application to the synthesis of 0231B in 10 steps (8.1% overall yield) from 6-methylindole 8 by introducing an acyl group into the 3-position of the indole nucleus, cyclization of the side chain at the 3-position to the 4-position and subsequent elimination of the phenyl group, and conjugate addition of the substituted phenyl group.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Methylindol, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.