Merck
Novel anti-diabetic effect of SCM-198 via inhibiting the hepatic NF-κB pathway in db/db mice.
Bioscience reports (2011-08-24)
Hui Huang, Hong Xin, Xinhua Liu, Yajun Xu, Danyi Wen, Yahua Zhang, Yi Zhun Zhu
ZUSAMMENFASSUNG
There are reports of early evidence that suggest the involvement of chronic low-grade inflammation in the pathogenesis of Type 2 diabetes. Thus, substances that have effects in reducing inflammation could be potential drugs for Type 2 diabetes. Leonurine (4-guanidino-n-butyl syringate; SCM-198) is an alkaloid in HL (Herba leonuri), which was reported to possess anti-inflammatory properties. We hypothesize that SCM-198 may have beneficial effects on Type 2 diabetes. In the present study, we attempted to test this hypothesis by evaluating the anti-diabetic effect of SCM-198 and the possible underlying mechanisms of its effects in db/db mice. SCM-198 (50, 100 and 200 mg/kg of body weight), pioglitazone (50 mg/kg of body weight, as a positive control) or 1% CMC-Na (sodium carboxymethylcellulose) were administered to the db/db or db/m mice once daily for 3 weeks. After 3 weeks, SCM-198 (200 mg/kg of body weight) treatment significantly reduced the fasting blood glucose level and increased the plasma insulin concentration in the db/db mice, meanwhile it significantly lowered the plasma TAG (triacylglycerol) concentration and increased the HDL (high-density lipoprotein)-cholesterol concentration. Moreover, the dysregulated transcription of the hepatic glucose metabolic enzymes, including GK (glucokinase), G6Pase (glucose-6-phosphatase) and PEPCK (phosphoenolpyruvate carboxykinase), was recovered by an Akt-dependent pathway. The pro-inflammatory mediators {such as TNFα (tumour necrosis factor α), IL (interleukin)-6, IL-1β, degradation of IκB [inhibitor of NF-κB (nuclear factor-κB)] α and thereafter activation of NF-κB} were reversed by SCM-198 treatment in the db/db mice. The present study provides first evidence that SCM-198 exhibits anti-inflammatory activity and has an ameliorating effect on diabetic symptoms via inhibiting of NF-κB/IKK (IκB kinase) pathway. Consequently, we suggest that SCM-198 may be a prospective agent for prevention and/or moderation of the progress of Type 2 diabetes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Leonurine, ≥98% (HPLC)