Merck
  • Proton efflux coupled to dark H2 oxidation in whole cells of a marine sulfur photosynthetic bacterium (Chromatium sp. strain Miami PBS1071).
Proton efflux coupled to dark H2 oxidation in whole cells of a marine sulfur photosynthetic bacterium (Chromatium sp. strain Miami PBS1071).
Journal of bacteriology (1983-04-01)
S Kumazawa, S Izawa, A Mitsui
ZUSAMMENFASSUNG
Whole cells of photoanaerobically grown Chromatium sp. strain Miami PBS1071, a marine sulfur purple bacterium, oxidized H2 in the dark through the oxyhydrogen reaction at rates of up to 59 nmol of H2 per mg (dry weight) per min. H2 oxidation was routinely measured in H2 pulse experiments with air-equilibrated cells. The reaction was accompanied by a reversible H+ efflux from the cells, suggesting an outward H+ translocation reaction coupled to H2 oxidation. The H+/e- ratio, calculated from simultaneous measurements of H2, O2, and H+ changes in the medium, varied with the cultures from 0.7 to 1.2. The ratio increased considerably when the backflow of H+ was taken into account. Anaerobic H2 uptake with 2,5-dimethyl-p-benzoguinone as an oxidant also showed a weak H+-translocating activity. No H+-translocating activity was detected with methylene blue as an oxidant. Carbonylcyanide 3-chlorophenylhydrazone (1 microM) stimulated H2 oxidation and abolished the associated H+ changes when H2 oxidation was observed in O2 pulse experiments with H2-Ar-equilibrated cells. However, the uncoupler inhibited both H2 oxidation and H+ changes when measurements were made in H2 pulse experiments with air-equilibrated cells. It is suggested that in this bacterium the susceptibility of hydrogenase to reversible O2 inactivation in situ is enhanced by the presence of uncoupling agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,5-Dimethyl-1,4-benzochinon, ≥98.0%