Merck
  • Fabrication of vascular endothelial growth factor antibody bioconjugated ultrasmall near-infrared fluorescent Ag2S quantum dots for targeted cancer imaging in vivo.
Fabrication of vascular endothelial growth factor antibody bioconjugated ultrasmall near-infrared fluorescent Ag2S quantum dots for targeted cancer imaging in vivo.
Chemical communications (Cambridge, England) (2013-03-19)
Yong Wang, Xiu-Ping Yan
ZUSAMMENFASSUNG
BSA-stabilized Ag2S QDs were fabricated in aqueous solution via a one-pot approach, and subsequently bioconjugated with antiVEGF for targeted in vivo cancer imaging.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Silbersulfid, 99.9% trace metals basis