Merck
  • Effects of monthly intramuscular neridronate in rheumatic patients in chronic treatment with low-dose glucocorticoids.
Effects of monthly intramuscular neridronate in rheumatic patients in chronic treatment with low-dose glucocorticoids.
Clinical and experimental rheumatology (2009-09-24)
M Benucci, G Saviola, P Baiardi, L Abdi-Ali, M R Povino, S Dolenti, L Campostrini, S Sacco, M Manfredi, M Rossini
ZUSAMMENFASSUNG
To assess the effects of intramuscular (im) neridronate (NE) on lumbar and femoral neck BMD and on markers of bone turnover in rheumatic patients under chronic low-dose glucocorticoids (GC) therapy. Sixty-nine osteopoenic and osteoporotic patients, affected by rheumatic diseases and gastric or esophageal conditions which contraindicated treatment with oral bisphosphonates (BPs), were randomly assigned to: Group A (23 patients) administered with daily calcium 1 g and vitamin D 800 UI; Group B (46 patients) receiving daily calcium 1 g, vitamin D 800 UI and im NE 25 mg monthly. After 12 months of therapy (M12) lumbar BMD was reduced of 2.97% in Group A, and improved of 3.34% (p=0.001) in Group B; at M12, femoral neck BMD was reduced of 2.40% in Group A and improved of 1.78% in Group B (p=0.010). After 6 (M6) and 12 months of therapy, the bone resorption markers were significantly reduced in Group B: OHPr-41.64% at M6 (p<0.001) and -37.91% at M12 (p<0.001); DPD-33.4% at M6 (p<0.001) and -33.18% (p<0.001) at M12: NTX -57.08% (p<0.001) at M6 and -55.95% (p<0.001) at M12; OC-11.62% (p=0.05) at M6 and -12.62% at M12 (p=0.06); B-ALP -13.95 % at M6 (p=0.04) and -0.85% at M12 (NS). A twelve-month intramuscular NE treatment in rheumatic patients under GCs therapy improves lumbar and femoral BMD and mainly reduces the markers of bone resorption.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Neridronate, ≥97% (NMR), solid