Merck
Synthesis of beta-(S-methyl)thioaspartic acid and derivatives.
Bioorganic & medicinal chemistry (2008-05-13)
Jorge Heredia-Moya, Kenneth L Kirk
ZUSAMMENFASSUNG
Beta-(S-Methyl)thioaspartic acid occurs as a posttranslational modification at position 88 in Escherichia coli ribosomal protein S12, a position that is a mutational hotspot resulting in both antibiotic-resistant and antibiotic-sensitive phenotypes. Critical to research designed to determine the biological function of beta-(S-methyl)thioaspartic acid will be the availability of synthetic beta-(S-methyl)thioaspartic acid as well as derivatives designed for peptide incorporation. We report here the synthesis of beta-(S-methyl)thioaspartic acid and derivatives. The installation of the beta-methylthio moiety into the aspartic acid structure was accomplished by electrophilic sulfenylation of N-protected-l-aspartic acid derivatives with 2,4-dinitrophenyl methyl disulfide. Following this key transformation, we were able to prepare protected beta-(S-methyl)thioaspartic acid derivative suitable for peptide coupling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-threo-β-Methylaspartic acid