Merck
  • Probing the mechanism of the carcinogenic activation of N-nitrosodiethanolamine with deuterium isotope effects: in vivo induction of DNA single-strand breaks and related in vitro assays.
Probing the mechanism of the carcinogenic activation of N-nitrosodiethanolamine with deuterium isotope effects: in vivo induction of DNA single-strand breaks and related in vitro assays.
Chemical research in toxicology (1998-12-22)
R N Loeppky, A Fuchs, C Janzowski, C Humberd, P Goelzer, H Schneider, G Eisenbrand
ZUSAMMENFASSUNG
A series of bioassays, including in vivo induction of DNA single-strand breaks (SSB) and cytotoxicity in cytochrome P450 2E1-transfected cells, were utilized with N-nitrosodiethanolamine (NDELA), its deuterated isotopomers (alpha-D4NDELA and beta-D4NDELA), N-nitroso-2-hydroxymorpholine (NHMOR), and two of its deuterated isotopomers (2-D-NHMOR and 5,5-D2-NHMOR) to probe the mechanism of carcinogenic activation of NDELA and the role of its metabolite NHMOR. DNA samples, taken from the livers of male Wistar rats 4 h after the administration of NDELA, exhibited dose-dependent DNA SSB levels over the range of 0.08-0.75 mmol/kg (body weight), with the greatest SSB level at the highest dose. Deuterium isotope effects on DNA SSB levels were inversely dependent on dose: alpha-D4NDELA, 3. 22-1.37; and beta-D4NDELA, 1.38-0.79. At the lowest dose of 0.15 mmol/kg (body weight), 5,5-D2-NHMOR gave an isotope effect for DNA SSB of 2.8 while that for 2-D-NHMOR was 0.7. NDELA and beta-D4NDELA were equally cytotoxic to human P450 2E1-transfected V79 Chinese hamster cells, while alpha-D4NDELA was not. Significant DNA SSB levels were observed in these cells for NDELA and beta-D4NDELA but not for alpha-D4NDELA. A kinetic deuterium isotope effect of 2.6 for Vmax/Km was observed for the horse liver alcohol dehydrogenase-mediated oxidation of beta-D4NDELA to NHMOR, while kH/kD for alpha-D4NDELA was 1.05. These data provide the first definitive evidence for the activation of NDELA by a pathway involving the scission of the alpha-CH bond and are consistent with P450 2E1-mediated alpha-hydroxylation of NDELA producing the corresponding reactive alpha-hydroxynitrosamine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Nitrosodiethanolamin