Merck
  • 3,4-Diaminopyridine improves neuromuscular transmission in a MuSK antibody-induced mouse model of myasthenia gravis.
3,4-Diaminopyridine improves neuromuscular transmission in a MuSK antibody-induced mouse model of myasthenia gravis.
Journal of neuroimmunology (2012-03-14)
Shuuichi Mori, Masahiko Kishi, Sachiho Kubo, Takuyu Akiyoshi, Shigeru Yamada, Tsuyoshi Miyazaki, Tetsuro Konishi, Naoki Maruyama, Kazuhiro Shigemoto
ZUSAMMENFASSUNG
This study investigated the effect of 3,4-diaminopyridine (3,4-DAP), a potent potentiator of transmitter release, on neuromuscular transmission in vivo in a mouse model of myasthenia gravis (MG) caused by antibodies against muscle-specific kinase (MuSK; MuSK-MG) and ex vivo in diaphragm muscle from these mice. 3,4-DAP significantly improved neuromuscular transmission, predominantly by increasing acetylcholine (ACh) release, supporting presynaptic potentiation as an effective treatment strategy for MuSK-MG patients who have defective transmitter release. In MuSK-MG, we suggest that only low-dose acetylcholinesterase (AChE) inhibitors be used to avoid side effects, and we propose that 3,4-DAP may be effective as a symptomatic therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,4-Diamino-pyridin, ≥98%